6-Propyl-2-thiouracil

Warning

Synonyms: Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

P3755-100G P3755-25G P3755-10G
220203-404EA 306.92 USD
220203-404 220203-406 220203-405
6-Propyl-2-thiouracil
6-Propyl-2-thiouracil
Formula: C₇H₁₀N₂OS
MW: 170.24 g/mol
Boiling Pt: 448 °C (1013 hPa)
Melting Pt: 215 - 218 °C
MDL Number: MFCD00006041
CAS Number: 51-52-5
EINECS: 200-103-2
Merck Index: 12,08054

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR