Diphenyl sulfoxide ≥96%

Synonyms: Phenyl sulphoxide, sulphinyldibenzene

P35405-100G P35405-25G
220202-918EA 112 USD
220202-918 220202-919
Diphenyl sulfoxide ≥96%
Diphenyl sulphoxide
Formula: C₁₂H₁₀OS
MW: 202.28 g/mol
MDL Number: MFCD00002085
CAS Number: 945-51-7
EINECS: 213-415-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR