Diphenyl sulfide ≥98%

Warning

Synonyms: Diphenylsulphane

P35316-25G P35316-100G
220202-913EA 43.62 USD
220202-913 220202-912
Diphenyl sulfide ≥98%
Diphenyl sulphide
Formula: C₁₂H₁₀S
MW: 186.28 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: -40 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003064
CAS Number: 139-66-2
EINECS: 205-371-4
UN: 2810
ADR: 6.1,III
Merck Index: 12,07467

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR