Polyethylene glycol 1,000

Synonyms: PEG 1,000, PEG 900-1100

P3515-1KG P3515-250G P3515-500G
220203-328EA 97.26 USD
220203-328 220203-329 220203-330
Polyethylene glycol 1,000
Polyethylene glycol 1,000
Formula: H(OCH₂CH₂)nOH
MW: 1000 g/mol
Boiling Pt: ? 250 °C (1013 hPa)
Melting Pt: 33 - 40 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 13,07651

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR