n-Pentadecane ≥99%

Danger

Synonyms: N-Pentadecane, Pentadecane

P3406-500G P3406-100G P3406-25G
220201-990EA 294 USD
220201-990 220201-988 220201-989
n-Pentadecane ≥99%
n-Pentadecane
Formula: H₃C(CH₂)₁₃CH₃
MW: 212.42 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 13…15 °C
Density: 0.768 g/cm³ (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00008990
CAS Number: 629-62-9
EINECS: 211-098-1
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR