4-Phenylpyridine

Warning

P33429-5G P33429-25G
220203-290EA 97.14 USD
220203-290 220203-289
4-Phenylpyridine
4-Phenylpyridine
Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00006420
CAS Number: 939-23-1
EINECS: 213-357-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR