2-Phenylpyridine

Warning

P33402-50G P33402-10G
220203-288EA 439.38 USD
220203-288 220203-287
2-Phenylpyridine
2-Phenylpyridine
Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 269…270 °C (1013 hPa)
Melting Pt: 76…77 °C
Density: 1.623 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5
EINECS: 213-763-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR