Polyethylene glycol 400

Synonyms: PEG 400

P3265-500G P3265-1KG
220203-260EA 69.05 USD
220203-260 220203-259
Polyethylene glycol 400
Polyethylene glycol 400
Formula: H(OCH₂CH₂)nOH
MW: 400 g/mol
Boiling Pt: ? 250 °C (1013 hPa)
Melting Pt: 4 - 8 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR