3-Phenylpropylamine ≥98%

Synonyms: 3-Phenyl-1-propylamine

P32406-10G P32406-25G
220203-248EA 65.69 USD
220203-248 220203-249
3-Phenylpropylamine ≥98%
3-Phenylpropylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 221 °C (1013 hPa)
Density: 0.951 g/cm³ (15 °C)
MDL Number: MFCD00008224
CAS Number: 2038-57-5
EINECS: 218-012-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR