2-Phenylpropionic acid ≥97%

Danger

Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid

P31701-10G P31701-5G
220203-232EA 83.14 USD
220203-232 220203-233
2-Phenylpropionic acid ≥97%
2-Phenylpropionic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: –5 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002650
CAS Number: 492-37-5
EINECS: 207-752-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR