Propyl-3,4,5-trihydroxybenzoate

Warning

Synonyms: Gallic acid propyl ester, Propyl gallate, Propyl 3,4,5-trihydroxybenzoate, Progalline P, N-Propyl gallate

P3130-500G P3130-100G
220203-216EA 364.06 USD
220203-216 220203-215
Propyl-3,4,5-trihydroxybenzoate
Propyl-3,4,5-trihydroxybenzoate
Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 449 °C (1013 hPa)
Melting Pt: 145 - 148 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002196
CAS Number: 121-79-9
EINECS: 204-498-2
Merck Index: 12,08044

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR