2-Phenyl-2-propanol ≥97%

Warning

Synonyms: α,α-Dimethylbenzyl alcohol

P30802-25G P30802-5G
220203-186EA 106.29 USD
220203-186 220203-187
2-Phenyl-2-propanol ≥97%
2-Phenyl-2-propanol
Formula: C₆H₅C(OH)(CH₃)₂
MW: 136.19 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: 33…35 °C
Density: 0.973 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004456
CAS Number: 617-94-7
EINECS: 210-539-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f8edbb976395240a99a4f52addfbfe32e8c926f&vid=5b4ae0c8b87b317b3d1326768c6ff8bdaaab37f6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR