Polyethylene glycol 200

Synonyms: PEG 200, Ethylene glycol polymer average Mn 200

P3015-500G P3015-1KG P3015-250G P3015-20KG P3015-5G
220203-157EA 46.23 USD
220203-157 220203-154 220203-156 220203-155 220203-158
Polyethylene glycol 200
Polyethylene glycol 200
Formula: H(OCH₂CH₂)nOH
MW: 200 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: –55…–40 °C
Density: 1.124 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD01779596
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR