1-Phenylpiperazine

Synonyms: N-Phenylpiperazine

P30004-100G P30004-25G
220203-146EA 130.31 USD
220203-146 220203-147
1-Phenylpiperazine
1-Phenylpiperazine
Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 17 - 19 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005957
CAS Number: 92-54-6
EINECS: 202-165-6
UN: 2927
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR