N-Phenylmaleimide

P27100-100G P27100-25G
220202-630EA 99.51 USD
220202-630 220202-631
N-Phenylmaleimide
N-Phenylmaleimide
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 84 - 86 °C
MDL Number: MFCD00005502
CAS Number: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,07452

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR