m-Phenylenediamine ≥99%

Danger

Synonyms: MPD, 1,3-Diaminobenzene, 1,3-Phenylenediamine

P23954-15KG P23954-100G P23954-1KG P23954-5G P23954-500G
220202-975EA 687.08 USD
220202-975 220202-974 220202-976 220202-978 220202-977
m-Phenylenediamine ≥99%
m-Phenylenediamine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 63…64 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 187 °C
Storage Temperature: Ambient
MDL Number: MFCD00007799
CAS Number: 108-45-2
EINECS: 203-584-7
UN: 1673
ADR: 6.1,III
Merck Index: 12,07437

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR