Tween® 20 (Polysorbate)

Synonyms: Polyoxyethylene sorbitan monolaurate

P2287-100ML P2287-500ML
220200-359EA 52.32 USD
220200-359 220200-361
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR