Cyclohexen-1-ylbenzene ≥95%

Synonyms: 1-Phenylcyclohexene, 1-Phenyl-1-cyclohexene, 2,3,4,5-Tetrahydro-1,1'-biphenyl

P22303-5G P22303-25G
220200-181EA 73.77 USD
220200-181 220200-180
Cyclohexen-1-ylbenzene ≥95%
Cyclohexen-1-ylbenzene
Formula: C₁₂H₁₄
MW: 158.24 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.004 g/cm³ (0 °C)
MDL Number: MFCD00001542
CAS Number: 771-98-2
EINECS: 212-242-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce31989847b7f5823ac4c86db68428fc148afb1d&vid=eeeb1ffa4c17dd4e58f5fc1f819153e435054e02&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR