(±)-2-Phenylbutyric acid ≥98%

Synonyms: (±)-2-Phenylbutanoic acid, 2-Phenylbutanoic acid

P20955-500G P20955-100G
220202-781EA 50.08 USD
220202-781 220202-780
(±)-2-Phenylbutyric acid ≥98%
(±)-2-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 39…42 °C
MDL Number: MFCD00002667
CAS Number: 90-27-7
EINECS: 201-982-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR