Phenylboronic acid ≥95%

Warning

Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid

P20009-250G P20009-10G P20009-50G
220202-740EA 193.85 USD
220202-740 220202-739 220202-741
Phenylboronic acid ≥95%
Phenylboronic acid
Formula: C₆H₇BO₂
MW: 121.93 g/mol
Melting Pt: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002103
CAS Number: 98-80-6
EINECS: 202-701-9
Merck Index: 13,01068

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR