Sodium propionate ≥99.0%

Warning

Synonyms: Propionic acid sodium salt, Sodiumpropionate

P1880-1KG P1880-100G P1880-500G
220202-700EA 95.63 USD
220202-700 220202-699 220202-701
Sodium propionate ≥99.0%
Sodium propionate
Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 288.76 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002759
CAS Number: 137-40-6
EINECS: 205-290-4
Merck Index: 12,08816

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR