D-α-Phenylalanine ≥98%

Synonyms: D-Phenylalanine, H-D-Phe-OH, 3-Phenyl-D-alanine, 3-Phenyl-D(+)-alanine

P1751-100G P1751-5G P1751-25G
220201-693EA 428.31 USD
220201-693 220201-695 220201-694
D-α-Phenylalanine ≥98%
D-α-Phenylalanine
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 295 °C (1013 hPa)
MDL Number: MFCD00004270
CAS Number: 673-06-3
EINECS: 211-603-5
Merck Index: 12,07425

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR