Phenylacetyl chloride ≥98%

P16753-5G P16753-100G P16753-500G
220202-606EA 110.92 USD
220202-606 220202-604 220202-605
Phenylacetyl chloride ≥98%
Phenylacetyl chloride
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 91 °C
Density: 1.1682 g/cm³ (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00000729
CAS Number: 103-80-0
EINECS: 203-146-5
UN: 2577
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR