Tween® 60 (Polysorbate)

Synonyms: Polyoxyethylene sorbitan monostearate

P1629-500ML P1629-100ML P1629-1GA
220202-572EA 89.34 USD
220202-572 220202-570 220202-571
Tween® 60 (Polysorbate)
Tween® 60 (Polysorbate)
Formula: C₆₄H₁₂₆O₂₆
Boiling Pt: ∼110 °C (1013 hPa)
Density: 1.08 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD01780110
CAS Number: 9005-67-8
EINECS: 500-020-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR