Pararosaniline hydrochloride, certified by the Biological Stain Commission

Danger

Synonyms: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride

P1528-25G P1528-100G
220202-531EA 438.15 USD
220202-531 220202-530
Pararosaniline hydrochloride, certified by the Biological Stain Commission
Pararosaniline hydrochloride
Formula: C₁₉H₁₈ClN₃
MW: 323.82 g/mol
Melting Pt: 268 °C
Storage Temperature: Ambient
MDL Number: MFCD00001657
CAS Number: 569-61-9
EINECS: 209-321-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR