trans-Platin

Synonyms: trans-Diamminedichloroplatinum (II), trans-Diammineplatinum (II) dichloride, trans-Dichlorodiammine platinum(II), trans-Dichlorodiamineplatinum(II)

P1525-1G P1525-250MG
220202-521EA 473.85 USD
220202-521 220202-522
trans-Platin
trans-Platin
Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 340 °C
Density: 3.746 g/cm³ (20 °C)
MDL Number: MFCD00011623
CAS Number: 14913-33-8
EINECS: 238-980-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR