Perylene

P11204-1G P11204-5G
220202-361EA 88.34 USD
220202-361 220202-362
Perylene
Perylene
Formula: C₂₀H₁₂
MW: 252.32 g/mol
Boiling Pt: 443 °C (1013 hPa)
Melting Pt: 273…278 °C
Density: 1.362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004142
CAS Number: 198-55-0
EINECS: 205-900-9
Merck Index: 12,07325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fecd171289d227be926b874d4a5001d67e77fe55&vid=6ff19028cb9ccd06fd2f57b58d5a140093f9ced8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR