Potassium tellurite ≥90%, Sigma-Aldrich®

Danger

Synonyms: Potassium tellurite, Potassium tellurium trioxide, Potassium tellurium (IV) oxide

P0677-100G P0677-25G
220201-473EA 403.08 USD
220201-473 220201-474
Potassium tellurite ≥90%, Sigma-Aldrich®
Potassium tellurite
Formula: K₂TeO₃
MW: 253.79 g/mol
MDL Number: MFCD00011379
CAS Number: 7790-58-1
EINECS: 232-213-1
UN: 3284
ADR: 6.1,II

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR