Palmitic acid ≥99%

Synonyms: Hexadecanoic acid, Hexadecylic acid

P0500-100G P0500-250G P0500-25G P0500-10G
220202-099EA 465.23 USD
220202-099 220202-101 220202-102 220202-100
Palmitic acid ≥99%
Palmitic acid
Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 61…63 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR