n-Octadecane ≥99%

Synonyms: Octadecane

O652-100G O652-500G O652-25G
220201-616EA 82.06 USD
220201-616 220201-618 220201-617
n-Octadecane ≥99%
n-Octadecane
Formula: H₃C(CH₂)₁₆CH₃
MW: 254.5 g/mol
Boiling Pt: 316.1 °C (1013 hPa)
Melting Pt: 26…29 °C
Density: 0.7765 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00009007
CAS Number: 593-45-3
EINECS: 209-790-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR