1,1,3,3-Tetramethylbutylamine ≥95%

Synonyms: 2,4,4-Trimethylpentan-2-amine, 2-Amino-2,4,4-trimethylpentane, tert-Octylamine

O6000-100G O6000-500G
220201-587EA 58.15 USD
220201-587 220201-588
1,1,3,3-Tetramethylbutylamine ≥95%
1,1,3,3-Tetramethylbutylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 137 °C (1013 hPa)
Melting Pt: –8 °C
Density: 0.805 g/cm³ (20 °C)
MDL Number: MFCD00008053
CAS Number: 107-45-9
EINECS: 203-491-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR