Octanoyl chloride ≥99%

O4733-500ML O4733-100ML O4733-5ML
220201-532EA 86.69 USD
220201-532 220201-531 220201-533
Octanoyl chloride ≥99%
Octanoyl chloride
Formula: C₈H₁₅ClO
MW: 162.66 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000766
CAS Number: 111-64-8
EINECS: 203-891-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR