Orange G

Synonyms: CI 16230

O3756-100G O3756-25G
220201-352EA 233.85 USD
220201-352 220201-353
Orange G
Orange G
Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Melting Pt: 141 °C
Density: 0.45 to 0.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR