1,8-Octanedithiol ≥97%

Warning

Synonyms: 1,8-Dimercaptooctane

O3605-5G O3605-1G
220201-344EA 198.77 USD
220201-344 220201-343
1,8-Octanedithiol ≥97%
1,8-Octanedithiol
Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 270 °C (1013 hPa)
Density: 0.9814 g/cm³ (0 °C)
MDL Number: MFCD00003574
CAS Number: 1191-62-4
EINECS: 214-738-8
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR