Octadecyl isocyanate, Technical Grade

Synonyms: Stearyl isocyanate, Isocyanic acid octadecyl ester, Isocyanic acid stearyl ester

O1807-500G O1807-100G
220201-268EA 249.85 USD
220201-268 220201-267
Octadecyl isocyanate, Technical Grade
Octadecyl isocyanate
Formula: C₁₉H₃₇NO
MW: 295.51 g/mol
Boiling Pt: 140…141 °C (0.4 mmHg)
Melting Pt: 15…16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002041
CAS Number: 112-96-9
EINECS: 204-019-7
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR