Octadecylamine ≥90%, Technical Grade

Danger

Synonyms: Octadecan-1-amine, 1-Octadecylamine, Stearylamine, 1-Aminooctadecane

O1408-500G O1408-3KG
220201-260EA 57.51 USD
220201-260 220201-259
Octadecylamine ≥90%, Technical Grade
Octadecylamine
Formula: H₃C(CH₂)₁₆CH₂NH₂
MW: 269.51 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 50 - 52 °C
Density: 0.8618 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008159
CAS Number: 124-30-1
EINECS: 204-695-3
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR