4'-Nitroacetophenone ≥98%

Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone

N9608-100G N9608-5G
220201-042EA 73.77 USD
220201-042 220201-043
4'-Nitroacetophenone ≥98%
4'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 76 - 78 °C
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007355
CAS Number: 100-19-6
EINECS: 202-827-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR