3'-Nitroacetophenone ≥99%

Synonyms: m-Nitroacetophenone

N9381-500G N9381-100G
220201-030EA 133.54 USD
220201-030 220201-029
3'-Nitroacetophenone ≥99%
3'-Nitroacetophenone
Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 77 - 78 °C
Density: 1.433 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007259
CAS Number: 121-89-1
EINECS: 204-504-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR