N-(1-Naphthyl)ethylenediamine dihydrochloride

Warning

N9125-25G N9125-100G N9125-10G
220201-003EA 128.15 USD
220201-003 220201-001 220201-002
N-(1-Naphthyl)ethylenediamine dihydrochloride
N-(1-Naphthyl)ethylenediamine dihydrochloride
Formula: C₁₂H₁₆Cl₂N₂
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR