1-Naphthylamine

Danger

Synonyms: 1-Aminonaphthalene

N9005-250G N9005-100G N9005-25G
220200-990EA 226.15 USD
220200-990 220200-989 220200-991
1-Naphthylamine
1-Naphthylamine
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 50 °C
Density: 1.171 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00004016
CAS Number: 134-32-7
EINECS: 205-138-7
UN: 2077
ADR: 6.1,III
Merck Index: 12,06485

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR