1-Naphthyl acetate ≥98%

Synonyms: Naphthalen-1-yl acetate, Ethanoic acid 1-naphthyl ester, Acetic acid 1-naphthyl ester, α-Naphthyl acetate, Acetic acid α-naphthyl ester, Ethanoic acid α-naphthyl ester

N8505-5G N8505-25G N8505-100G
220200-399EA 84.57 USD
220200-399 220200-398 220200-397
1-Naphthyl acetate ≥98%
1-Naphthyl acetate
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.1305 g/cm³ (50 °C)
Flash Pt: >100 °C
Storage Temperature: Freezer
MDL Number: MFCD00003922
CAS Number: 830-81-9
EINECS: 212-599-8

Order Now

Specification Test Results

Assay 98% ©

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR