L-Norvaline

Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid

N7627-10G N7627-1G N7627-5G
220200-161EA 289.23 USD
220200-161 220200-162 220200-163
L-Norvaline
L-Norvaline
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 101 °C
MDL Number: MFCD00064421
CAS Number: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR