Nitro blue tetrazolium chloride (NBT)

Warning

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

N6876-50MG N6876-5G N6876-100MG N6876-1G N6876-250MG N6876-10X10MG N6876-500MG
220200-756EA 59.22 USD
220200-756 220200-757 220200-751 220200-753 220200-754 220200-752 220200-755
Nitro blue tetrazolium chloride (NBT)
Nitro blue tetrazolium chloride (NBT)
Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de2c2cec7b7e51c50562030878bfa557af5df16&vid=175a4c932ab1f09b9b2613c727f5a3dadf3be4b0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR