Nickel (II) sulfate hexahydrate, ReagentPlus®

Danger

N4882-250G N4882-1KG
220200-529EA 81.23 USD
220200-529 220200-528
Nickel (II) sulfate hexahydrate, ReagentPlus®
Nickel(II) sulphate hexahydrate
Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3288
ADR: 6.1,III
Merck Index: 13,06541

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR