Nicotinic acid (Niacin)

Warning

N4126-5G N4126-500G N4126-1KG N4126-100G
220200-089EA 32.62 USD
220200-089 220200-088 220200-087 220200-454
Nicotinic acid (Niacin)
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR