2-Naphthylacetic acid

Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid

N4002-25G-A N4002-5G-A
220199-915EA 110.92 USD
220199-915 220199-916
2-Naphthylacetic acid
2-Naphthylacetic acid
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 139 - 142 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00004126
CAS Number: 581-96-4
EINECS: 209-475-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR