Nicotinamide

Warning

Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3

N3376-500G N3376-1KG N3376-100G
220200-275EA 99.45 USD
220200-275 220200-274 220200-273
Nicotinamide
Nicotinamide
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 127…128 °C
Density: 1.4463 g/cm³ (20 °C)
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR