beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98%

Synonyms: NADP sodium hydrate, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium monosodium salt xhydrate

N3139-1G N3139-100MG N3139-250MG
220200-242EA 763.2 USD
220200-242 220200-241 220200-243
beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98%
beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate
Formula: C₂₁H₂₇N₇NaO₁₇P₃·xH₂O
Melting Pt: 175 to 178 °C
MDL Number: MFCD00150480
CAS Number: 698999-85-8
EINECS: 214-664-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR