1-Nonene ≥96%

N30404-25G N30404-5G
220199-773EA 416.77 USD
220199-773 220199-774
1-Nonene ≥96%
1-Nonene
Formula: C₉H₁₈
MW: 126.24 g/mol
Boiling Pt: 147 °C (1013 hPa)
Melting Pt: –88 °C
Density: 0.7405 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009568
CAS Number: 124-11-8
EINECS: 204-681-7
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR