β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate

Synonyms: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD

N3014-100MG N3014-5G N3014-1G N3014-10G
220200-220EA 52.62 USD
220200-220 220200-223 220200-222 220200-221
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
Formula: C₂₁H₂₇N₇O₁₄P₂·H₂O
Melting Pt: 140 to 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4
Merck Index: 13,06370

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR